CAS RN 89-68-9
CAS RN 89-68-9
IUPAC Name:
4-chloro-5-methyl-2-(propan-2-yl)phenol
Synonyms:
chlorothymol epa pesticide chemical code 080403 caswell no. 216 C10H13ClO 氯麝酚 caniprevent [veterinary] pioral
Molecular Weight (g/mol):
184.66
Molecular Formula:
C10H13ClO
InChi Key:
KFZXVMNBUMVKLN-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC(Cl)=C(C)C=C1O
1
–
1
of
1
results
1
4-Chloro-2-isopropyl-5-methylphenol, 99%
CAS: 89-68-9 Molecular Formula: C10H13ClO Molecular Weight (g/mol): 184.66 MDL Number: MFCD00002326 InChI Key: KFZXVMNBUMVKLN-UHFFFAOYSA-N Synonym: chlorothymol,6-chlorothymol,4-chloro-2-isopropyl-5-methylphenol,chlorthymol,4-chlorothymol,thymol, 6-chloro,phenol, 4-chloro-5-methyl-2-1-methylethyl,caswell no. 216,4-chloro-6-isopropyl-3-methylphenol,unii-lj25ti0cvt PubChem CID: 6982 IUPAC Name: 4-chloro-5-methyl-2-propan-2-ylphenol SMILES: CC(C)C1=CC(Cl)=C(C)C=C1O