accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 89-59-8

CH3ClNOO

CAS RN 89-59-8

IUPAC Name: 4-chloro-1-methyl-2-nitrobenzene
Synonyms: 4-chloro-1-methyl-2-nitrobenzene
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: SQFLFRQWPBEDHM-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results