accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 888711-47-5

BIFFFK

CAS RN 888711-47-5

IUPAC Name: potassium trifluoro(iodomethyl)boranuide
Synonyms: potassium trifluoro(iodomethyl)boranuide
Molecular Weight (g/mol): 247.83
Molecular Formula: CH2BF3IK
InChi Key: WQJVWRYIXQXISQ-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)CI

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results