accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 886851-54-3

H3CHNSBrOO

CAS RN 886851-54-3

IUPAC Name: ({7-bromo-2H,3H-thieno[3,4-b][1,4]dioxin-5-yl}methyl)(methyl)amine
Synonyms: ({7-bromo-2H,3H-thieno[3,4-b][1,4]dioxin-5-yl}methyl)(methyl)amine
Molecular Weight (g/mol): 264.14
Molecular Formula: C8H10BrNO2S
InChi Key: FJHBRYOEMIQSPC-UHFFFAOYSA-N
SMILES: CNCC1=C2OCCOC2=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results