accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NHNN

CAS RN 886851-49-6

IUPAC Name: methyl({[3-(pyrimidin-2-yl)phenyl]methyl})amine
Synonyms: methyl({[3-(pyrimidin-2-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 199.26
Molecular Formula: C12H13N3
InChi Key: AHOVMAYXXRVWIS-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results