accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNNS

CAS RN 886851-41-8

IUPAC Name: methyl({[6-(thiophen-2-yl)pyridin-3-yl]methyl})amine
Synonyms: methyl({[6-(thiophen-2-yl)pyridin-3-yl]methyl})amine
Molecular Weight (g/mol): 204.29
Molecular Formula: C11H12N2S
InChi Key: LFZDJRPWILOVEO-UHFFFAOYSA-N
SMILES: CNCC1=CN=C(C=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results