accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNOOH

CAS RN 886851-33-8

IUPAC Name: [3-(furan-2-yl)-1-methyl-1H-pyrazol-5-yl]methanol
Synonyms: [5-(furan-2-yl)-2-methylpyrazol-3-yl]methanol
Molecular Weight (g/mol): 178.19
Molecular Formula: C9H10N2O2
InChi Key: BYXYWFXMSJTOKB-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1CO)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results