accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 886761-73-5

CH3OOHOFF

CAS RN 886761-73-5

IUPAC Name: 2-(4,5-difluoro-2-methoxyphenyl)acetic acid
Synonyms: (4,5-difluoro-2-methoxyphenyl)acetic acid
Molecular Weight (g/mol): 202.16
Molecular Formula: C9H8F2O3
InChi Key: UYEGFRSSHGRVJF-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(F)C=C1CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results