CAS RN 886503-38-4
IUPAC Name:
4-(bromomethyl)-1-chloro-2-[(trifluoromethyl)sulfanyl]benzene
Synonyms:
4-(bromomethyl)-1-chloro-2-[(trifluoromethyl)sulfanyl]benzene
Molecular Weight (g/mol):
305.54
Molecular Formula:
C8H5BrClF3S
InChi Key:
QRIMMRPSWRLACM-UHFFFAOYSA-N
SMILES:
FC(F)(F)SC1=C(Cl)C=CC(CBr)=C1
1
–
1
of
1
results
1
4-Chloro-3-(trifluoromethylthio)benzyl bromide, 97%, Thermo Scientific™
CAS: 886503-38-4 Molecular Formula: C8H5BrClF3S Molecular Weight (g/mol): 305.537 MDL Number: MFCD06660314 InChI Key: QRIMMRPSWRLACM-UHFFFAOYSA-N Synonym: 4-chloro-3-trifluoromethylthio benzyl bromide,4-bromomethyl-1-chloro-2-trifluoromethyl sulfanyl benzene PubChem CID: 17750740 IUPAC Name: 4-(bromomethyl)-1-chloro-2-(trifluoromethylsulfanyl)benzene SMILES: C1=CC(=C(C=C1CBr)SC(F)(F)F)Cl