CAS RN 886500-75-0
IUPAC Name:
1-(2,3-difluoro-4-methoxyphenyl)methanamine
Synonyms:
1-(2,3-difluoro-4-methoxyphenyl)methanamine
Molecular Weight (g/mol):
173.16
Molecular Formula:
C8H9F2NO
InChi Key:
DBQHLJOOMYBKNQ-UHFFFAOYSA-N
SMILES:
COC1=C(F)C(F)=C(CN)C=C1
1
–
1
of
1
results
1
2,3-Difluoro-4-methoxybenzylamine, 97%, Thermo Scientific™
CAS: 886500-75-0 Molecular Formula: C8H9F2NO Molecular Weight (g/mol): 173.163 MDL Number: MFCD04115987 InChI Key: DBQHLJOOMYBKNQ-UHFFFAOYSA-N Synonym: 2,3-difluoro-4-methoxybenzylamine,2,3-difluoro-4-methoxyphenyl methanamine,2,3-difluoro-4-methoxy-phenyl methanamine,2,3-difluoro-4-methoxyphenyl methylamine,benzenemethanamine,2,3-difluoro-4-methoxy,2,3-bis fluoranyl-4-methoxy-phenyl methanamine,1-2,3-difluoro-4-methoxyphenyl methanamine PubChem CID: 3817603 IUPAC Name: (2,3-difluoro-4-methoxyphenyl)methanamine SMILES: COC1=C(C(=C(C=C1)CN)F)F