CAS RN 886499-58-7
IUPAC Name:
6-chloro-2-fluoro-3-methoxybenzoic acid
Synonyms:
6-chloro-2-fluoro-3-methoxybenzoic acid
Molecular Weight (g/mol):
204.58
Molecular Formula:
C8H6ClFO3
InChi Key:
LCBOXBUSYYCBLU-UHFFFAOYSA-N
SMILES:
COC1=C(F)C(C(O)=O)=C(Cl)C=C1
1
–
1
of
1
results
1
6-Chloro-2-fluoro-3-methoxybenzoic acid, 97%, Thermo Scientific™
CAS: 886499-58-7 Molecular Formula: C8H6ClFO3 Molecular Weight (g/mol): 204.58 MDL Number: MFCD04115947 InChI Key: LCBOXBUSYYCBLU-UHFFFAOYSA-N PubChem CID: 3856039 IUPAC Name: 6-chloro-2-fluoro-3-methoxybenzoic acid SMILES: COC1=C(F)C(C(O)=O)=C(Cl)C=C1