CAS RN 886499-04-3
IUPAC Name:
4-(bromomethyl)-2-fluoro-1-(trifluoromethoxy)benzene
Synonyms:
4-(bromomethyl)-2-fluoro-1-(trifluoromethoxy)benzene
Molecular Weight (g/mol):
273.03
Molecular Formula:
C8H5BrF4O
InChi Key:
MOWRYCSMYVKVNK-UHFFFAOYSA-N
SMILES:
FC1=C(OC(F)(F)F)C=CC(CBr)=C1
1
–
1
of
1
results
1
3-Fluoro-4-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™
CAS: 886499-04-3 Molecular Formula: C8H5BrF4O Molecular Weight (g/mol): 273.025 MDL Number: MFCD06660205 InChI Key: MOWRYCSMYVKVNK-UHFFFAOYSA-N Synonym: 3-fluoro-4-trifluoromethoxy benzyl bromide,4-bromomethyl-2-fluoro-1-trifluoromethoxy benzene,benzene,4-bromomethyl-2-fluoro-1-trifluoromethoxy PubChem CID: 17750672 IUPAC Name: 4-(bromomethyl)-2-fluoro-1-(trifluoromethoxy)benzene SMILES: C1=CC(=C(C=C1CBr)F)OC(F)(F)F