accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FOFFFO

CAS RN 886497-81-0

IUPAC Name: 2-fluoro-5-(trifluoromethoxy)benzaldehyde
Synonyms: 2-fluoro-5-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 208.11
Molecular Formula: C8H4F4O2
InChi Key: FMDGCHMPSCBYFX-UHFFFAOYSA-N
SMILES: FC1=CC=C(OC(F)(F)F)C=C1C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results