accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COClBrF

CAS RN 886497-36-5

IUPAC Name: 5-(bromomethyl)-1-chloro-3-fluoro-2-methoxybenzene
Synonyms: 5-(bromomethyl)-1-chloro-3-fluoro-2-methoxybenzene
Molecular Weight (g/mol): 253.5
Molecular Formula: C8H7BrClFO
InChi Key: SXXSRVKICKOIEA-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(CBr)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results