accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 886496-67-9

SH2NFFF

CAS RN 886496-67-9

IUPAC Name: 2-(trifluoromethyl)benzene-1-carbothioamide
Synonyms: 2-(trifluoromethyl)benzenecarbothioamide
Molecular Weight (g/mol): 205.2
Molecular Formula: C8H6F3NS
InChi Key: MBSSSIMORFRLOK-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results