accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 886496-49-7

OHOOFFFF

CAS RN 886496-49-7

IUPAC Name: 4-fluoro-3-(trifluoromethoxy)benzoic acid
Synonyms: 4-fluoro-3-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 224.11
Molecular Formula: C8H4F4O3
InChi Key: PTPKGMXRSREZFB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(OC(F)(F)F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results