accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 88536-11-2

H3CONH

CAS RN 88536-11-2

IUPAC Name: 4-propoxypiperidine
Synonyms: 4-propoxypiperidine
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: VOWMRECKIQVVPP-UHFFFAOYSA-N
SMILES: CCCOC1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results