CAS RN 885269-42-1
IUPAC Name:
4-{4'-methoxy-[1,1'-biphenyl]-4-sulfonamido}benzoic acid
Synonyms:
4-{4'-methoxy-[1,1'-biphenyl]-4-sulfonamido}benzoic acid
Molecular Weight (g/mol):
383.42
Molecular Formula:
C20H17NO5S
InChi Key:
ZFNQOUZSNZLJLR-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C1=CC=C(C=C1)S(=O)(=O)NC1=CC=C(C=C1)C(O)=O
1
–
1
of
1
results
1
4-(4'-Methoxy-4-biphenylylsulfonylamino)benzoic acid, 96%, Thermo Scientific™
CAS: 885269-42-1 Molecular Formula: C20H17NO5S Molecular Weight (g/mol): 383.418 MDL Number: MFCD06409458 InChI Key: ZFNQOUZSNZLJLR-UHFFFAOYSA-N Synonym: 4-4'-methoxy-1,1'-biphenyl-4-sulfonamido benzoic acid,4-4'-methoxy-1,1'-biphenyl-4-ylsulfonamido benzoic acid,4-4'-methoxy-4-biphenylylsulfonylamino benzoic acid PubChem CID: 44669168 IUPAC Name: 4-[[4-(4-methoxyphenyl)phenyl]sulfonylamino]benzoic acid SMILES: COC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)NC3=CC=C(C=C3)C(=O)O