accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClSOONHOHO

CAS RN 885269-32-9

IUPAC Name: 4-{4'-chloro-[1,1'-biphenyl]-4-sulfonamido}benzoic acid
Synonyms: 4-{4'-chloro-[1,1'-biphenyl]-4-sulfonamido}benzoic acid
Molecular Weight (g/mol): 387.83
Molecular Formula: C19H14ClNO4S
InChi Key: AYWSYJTWHIPSFL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(NS(=O)(=O)C2=CC=C(C=C2)C2=CC=C(Cl)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results