accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 884495-03-8

NFBrNH2

CAS RN 884495-03-8

IUPAC Name: 2-bromo-5-fluoropyridin-3-amine
Synonyms: 2-bromo-5-fluoropyridin-3-amine
Molecular Weight (g/mol): 191
Molecular Formula: C5H4BrFN2
InChi Key: QUZAKZBKMMUARE-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CN=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results