accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CON

CAS RN 88374-55-4

IUPAC Name: 4-(octyloxy)benzonitrile
Synonyms: 4-(octyloxy)benzonitrile
Molecular Weight (g/mol): 231.34
Molecular Formula: C15H21NO
InChi Key: GFNSBTARZPEIPN-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=C(C=C1)C#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results