accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 881913-20-8

BOHHO

CAS RN 881913-20-8

IUPAC Name: [3-(naphthalen-1-yl)phenyl]boronic acid
Synonyms: 3-(naphthalen-1-yl)phenylboronic acid
Molecular Weight (g/mol): 248.09
Molecular Formula: C16H13BO2
InChi Key: HFXYUCJLQZCNPD-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)C1=C2C=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results