accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOBrN

CAS RN 88139-91-7

IUPAC Name: (5-bromopyridin-2-yl)methanol
Synonyms: (5-bromopyridin-2-yl)methanol
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: RUCZFWMEACWFER-UHFFFAOYSA-N
SMILES: OCC1=CC=C(Br)C=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results