accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 879904-82-2

CH3H3CCH3OONSiH3CCH3HO

CAS RN 879904-82-2

IUPAC Name: tert-butyl 2-(hydroxydimethylsilyl)-1H-pyrrole-1-carboxylate
Synonyms: tert-butyl 2-(hydroxydimethylsilyl)pyrrole-1-carboxylate
Molecular Weight (g/mol): 241.36
Molecular Formula: C11H19NO3Si
InChi Key: RLDVDBYGGISJNL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=CC=C1[Si](C)(C)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results