accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 879896-62-5

CH3NHSNO

CAS RN 879896-62-5

IUPAC Name: methyl({[5-(morpholin-4-yl)thiophen-2-yl]methyl})amine
Synonyms: methyl({[5-(morpholin-4-yl)thiophen-2-yl]methyl})amine
Molecular Weight (g/mol): 212.31
Molecular Formula: C10H16N2OS
InChi Key: LTBKEKXCWIEXPM-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(S1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results