accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 879896-56-7

CH3NONNHH3C

CAS RN 879896-56-7

IUPAC Name: methyl({[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methyl})amine
Synonyms: methyl({[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 203.25
Molecular Formula: C11H13N3O
InChi Key: LJDBGVKOXDSIQQ-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC=C1C1=NC(C)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results