accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 879896-54-5

CH3NONO

CAS RN 879896-54-5

IUPAC Name: 2-(3-methyl-1,2,4-oxadiazol-5-yl)benzaldehyde
Synonyms: 2-(3-methyl-1,2,4-oxadiazol-5-yl)benzaldehyde
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: FPQLUUWDLSUKNS-UHFFFAOYSA-N
SMILES: CC1=NOC(=N1)C1=CC=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results