accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 879668-17-4

IUPAC Name: 3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine hydrochloride
Synonyms: 3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine hydrochloride
Molecular Weight (g/mol): 159.62
Molecular Formula: C6H10ClN3
InChi Key: CHLFJSVTUQAGSC-UHFFFAOYSA-N
SMILES: Cl.C1CC2=C(CN1)NC=N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results