CAS RN 87932-50-1
CAS RN 87932-50-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-3,4-dihydroxybenzoic acid, TRC
CAS: 87932-50-1 Molecular Formula: C7H5ClO4 Molecular Weight (g/mol): 188.56 IUPAC Name: 2-chloro-3,4-dihydroxybenzoic acid SMILES: OC(=O)c1ccc(O)c(O)c1Cl