accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 876316-47-1

IUPAC Name: 5-(furan-2-yl)-1-methyl-1H-pyrazole-3-carbonyl chloride
Synonyms: 5-(furan-2-yl)-1-methylpyrazole-3-carbonyl chloride
Molecular Weight (g/mol): 210.62
Molecular Formula: C9H7ClN2O2
InChi Key: RAYIOTXFHIKMIE-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C1=CC=CO1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results