accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 876316-44-8

IUPAC Name: 5-(bromomethyl)-2,4-diphenyl-1,3-thiazole
Synonyms: 5-(bromomethyl)-2,4-diphenyl-1,3-thiazole
Molecular Weight (g/mol): 330.24
Molecular Formula: C16H12BrNS
InChi Key: QDYJLVQRXQKPKL-UHFFFAOYSA-N
SMILES: BrCC1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results