accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOOH3CCH3H3CH3CO

CAS RN 876316-28-8

IUPAC Name: 2-[2-(furan-2-yl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[2-(furan-2-yl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 270.14
Molecular Formula: C16H19BO3
InChi Key: IKVHPKPYKOEGCK-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CC=C1C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results