accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 876-07-3

IUPAC Name: 4-(bromomethyl)benzoyl bromide
Synonyms: 4-(bromomethyl)benzoyl bromide
Molecular Weight (g/mol): 277.94
Molecular Formula: C8H6Br2O
InChi Key: VUYGJYAPDGKPBF-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(C=C1)C(Br)=O
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results