CAS RN 876-02-8
CAS RN 876-02-8
IUPAC Name:
1-(4-hydroxy-3-methylphenyl)ethan-1-one
Synonyms:
1-(4-hydroxy-3-methylphenyl)ethanone
Molecular Weight (g/mol):
150.18
Molecular Formula:
C9H10O2
InChi Key:
LXBHHIZIQVZGFN-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(O)C(C)=C1
1
–
1
of
1
results
1
4'-Hydroxy-3'-methylacetophenone, 98%
CAS: 876-02-8 Molecular Formula: C9H10O2 Molecular Weight (g/mol): 150.18 MDL Number: MFCD00002231 InChI Key: LXBHHIZIQVZGFN-UHFFFAOYSA-N Synonym: 4'-hydroxy-3'-methylacetophenone,4-hydroxy-3-methylacetophenone,1-4-hydroxy-3-methylphenyl ethanone,1-4-hydroxy-3-methylphenyl ethan-1-one,4-acetyl-2-methylphenol,ethanone, 1-4-hydroxy-3-methylphenyl,4-hydroxy-3-methyl acetophenone,1-4-hydroxy-3-methyl-phenyl-ethanone,1-acetyl-4-hydroxy-3-methylbenzene PubChem CID: 70135 SMILES: CC(=O)C1=CC=C(O)C(C)=C1