CAS RN 875664-57-6
CAS RN 875664-57-6
IUPAC Name:
4,5-difluoro-2-methylaniline
Synonyms:
4,5-difluoro-2-methylaniline
Molecular Weight (g/mol):
143.14
Molecular Formula:
C7H7F2N
InChi Key:
HKAMTWLPOCYTMX-UHFFFAOYSA-N
SMILES:
CC1=CC(F)=C(F)C=C1N
1
–
1
of
1
results
1
4,5-Difluoro-2-methylaniline, 99%, Thermo Scientific™
CAS: 875664-57-6 Molecular Formula: C7H7F2N Molecular Weight (g/mol): 143.14 MDL Number: MFCD08062378 InChI Key: HKAMTWLPOCYTMX-UHFFFAOYSA-N Synonym: 4,5-difluoro-2-methylbenzenamine,2-amino-4,5-difluorotoluene,4,5-difluoro-o-toluidine,2-amino-4,5-difluoro-toluene,4,5-difluoro-2-methylphenyl amine PubChem CID: 18727725 IUPAC Name: 4,5-difluoro-2-methylaniline SMILES: CC1=CC(F)=C(F)C=C1N