accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 87488-84-4

NNBr

CAS RN 87488-84-4

IUPAC Name: 1-(2-bromophenyl)-1H-pyrazole
Synonyms: 1-(2-bromophenyl)pyrazole
Molecular Weight (g/mol): 223.07
Molecular Formula: C9H7BrN2
InChi Key: QNDJHGODPWAKAO-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results