accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BNHOH3CCH3HOOH

CAS RN 874219-50-8

IUPAC Name: [4-(2-methylpropanamido)phenyl]boronic acid
Synonyms: 4-(2-methylpropanamido)phenylboronic acid
Molecular Weight (g/mol): 207.04
Molecular Formula: C10H14BNO3
InChi Key: DBWRRIVRDITVFV-UHFFFAOYSA-N
SMILES: CC(C)C(=O)NC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results