accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 874-66-8

CH3OO(Z)

CAS RN 874-66-8

IUPAC Name: (2Z)-3-(furan-2-yl)-2-methylprop-2-enal
Synonyms: (2Z)-3-(furan-2-yl)-2-methylprop-2-enal
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: ZNBXZUKDRRRQJK-ALCCZGGFSA-N
SMILES: C\C(C=O)=C\C1=CC=CO1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results