accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 872460-12-3

BFOOHHOOH

CAS RN 872460-12-3

IUPAC Name: 5-(dihydroxyboranyl)-2-fluorobenzoic acid
Synonyms: 5-(dihydroxyboranyl)-2-fluorobenzoic acid
Molecular Weight (g/mol): 183.93
Molecular Formula: C7H6BFO4
InChi Key: DGORTXQWDDXSIQ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(C(O)=O)=C(F)C=C1
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)