accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CSCH3

CAS RN 872-10-6

IUPAC Name: 1-(pentylsulfanyl)pentane
Synonyms: pentane, 1,1'-thiobis-
Molecular Weight (g/mol): 174.35
Molecular Formula: C10H22S
InChi Key: JOZDADPMWLVEJK-UHFFFAOYSA-N
SMILES: CCCCCSCCCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results