accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 871231-44-6

BOFFFK

CAS RN 871231-44-6

IUPAC Name: potassium trifluoro(3-formylphenyl)boranuide
Synonyms: potassium trifluoro(3-formylphenyl)boranuide
Molecular Weight (g/mol): 212.02
Molecular Formula: C7H5BF3KO
InChi Key: ZUJWOVZZESBYHA-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC(C=O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results