accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 870774-25-7

BOHHO

CAS RN 870774-25-7

IUPAC Name: [4-(naphthalen-1-yl)phenyl]boronic acid
Synonyms: 4-(naphthalen-1-yl)phenylboronic acid
Molecular Weight (g/mol): 248.09
Molecular Formula: C16H13BO2
InChi Key: BQHVXFQXTOIMQM-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results