accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 87-56-9

OHOClClO(Z)

CAS RN 87-56-9

IUPAC Name: (2Z)-2,3-dichloro-4-oxobut-2-enoic acid
Synonyms: mucochloric acid
Molecular Weight (g/mol): 168.96
Molecular Formula: C4H2Cl2O3
InChi Key: LUMLZKVIXLWTCI-IHWYPQMZSA-N
SMILES: OC(=O)C(\Cl)=C(\Cl)C=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)