accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COH2NH3C

CAS RN 868755-44-6

IUPAC Name: 1-(2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl)methanamine
Synonyms: 1-(2,2-dimethyl-3H-1-benzofuran-7-yl)methanamine
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: BQEPNISHAAOSFF-UHFFFAOYSA-N
SMILES: CC1(C)CC2=C(O1)C(CN)=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results