accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OCH3OOHO

CAS RN 866823-73-6

IUPAC Name: 3'-{[(2-cyclopentyl-6,7-dimethyl-1-oxo-2,3-dihydro-1H-inden-5-yl)oxy]methyl}-[1,1'-biphenyl]-4-carboxylic acid
Synonyms: biphenylindanone A
Molecular Weight (g/mol): 454.57
Molecular Formula: C30H30O4
InChi Key: KMKBEESNZAPKMP-UHFFFAOYNA-N
SMILES: CC1=C2C(=O)C(CC2=CC(OCC2=CC=CC(=C2)C2=CC=C(C=C2)C(O)=O)=C1C)C1CCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results