accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 866089-28-3

H3CHNOH3C

CAS RN 866089-28-3

IUPAC Name: 2,2-dimethyl-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 2,2-dimethyl-3,4-dihydro-1,4-benzoxazine
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: WILBVUMFTFLAHJ-UHFFFAOYSA-N
SMILES: CC1(C)CNC2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results