accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OHNOHO

CAS RN 86454-13-9

IUPAC Name: 6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid
Synonyms: 2-methyl-6-oxo-1H-pyridine-4-carboxylic acid
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: NQXYVTIQDNOHFM-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(=O)N1)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results