accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONNNH2

CAS RN 864068-88-2

IUPAC Name: 1-[2-(morpholin-4-yl)pyridin-4-yl]methanamine
Synonyms: 1-[2-(morpholin-4-yl)pyridin-4-yl]methanamine
Molecular Weight (g/mol): 193.25
Molecular Formula: C10H15N3O
InChi Key: NHFVDBCQOZATNT-UHFFFAOYSA-N
SMILES: NCC1=CC(=NC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results