accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNOH

CAS RN 864068-80-4

IUPAC Name: [3-(1H-pyrazol-1-yl)phenyl]methanol
Synonyms: [3-(pyrazol-1-yl)phenyl]methanol
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: VINVOQJANISHSK-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC=C1)N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results