CAS RN 86256-47-5
CAS RN 86256-47-5
IUPAC Name:
[3-fluoro-5-(trifluoromethoxy)phenyl]methanol
Synonyms:
[3-fluoro-5-(trifluoromethoxy)phenyl]methanol
Molecular Weight (g/mol):
210.13
Molecular Formula:
C8H6F4O2
InChi Key:
WJTUZFGGSZDUKV-UHFFFAOYSA-N
SMILES:
OCC1=CC(OC(F)(F)F)=CC(F)=C1
1
–
1
of
1
results
1
3-Fluoro-5-(trifluoromethoxy)benzyl alcohol, 97%, Thermo Scientific™
CAS: 86256-47-5 Molecular Formula: C8H6F4O2 Molecular Weight (g/mol): 210.128 MDL Number: MFCD18783225 InChI Key: WJTUZFGGSZDUKV-UHFFFAOYSA-N Synonym: 3-fluoro-5-trifluoromethoxy benzyl alcohol,3-fluoro-5-trifluoromethoxy phenyl methanol PubChem CID: 15582619 IUPAC Name: [3-fluoro-5-(trifluoromethoxy)phenyl]methanol SMILES: C1=C(C=C(C=C1OC(F)(F)F)F)CO